missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC5AC Antibody (MUC5AC/7798R), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24093MFV450
This item is not returnable.
View return policy
Description
MUC5AC Monoclonal antibody specifically detects MUC5AC in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MUC5AC | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| gastric mucin, leB, lewis B blood group antigen, major airway glycoprotein, MUC5, mucin 5, subtypes A and C, tracheobronchial/gastric, mucin 5AC, oligomeric mucus/gel-forming, mucin 5AC, oligomeric mucus/gel-forming pseudogene, mucin-5 subtype AC, tracheobronchial, mucin-5AC, TBM, tracheobronchial mucin | |
| Recombinant full-length human MUC5AC protein | |
| 0.1 mL | |
| Cellular Markers, Extracellular Matrix, Infections (Virus Bacteria and Parasites), Signal Transduction | |
| 4586 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| MUC5AC/7798R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction