missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC5AC Mouse anti-Human, Clone: 58M1, Abnova™
Mouse monoclonal antibody raised against native human MUC5AC.
Brand: Abnova MAB13371.100ug
This item is not returnable.
View return policy
Specifications
| MUC5AC | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| MUC5 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 4586 | |
| Store at 4°C. | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| 58M1 | |
| Mouse monoclonal antibody raised against native human MUC5AC. | |
| In 10mM PBS (0.05% BSA, 0.05% sodium azide). | |
| MUC5AC | |
| M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b) patient. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction