Learn More
MUC5B Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | MUC5B |
|---|---|
| Concentration | 333 μg/mL |
| Applications | Immunohistochemistry, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16866005
|
Proteintech
28118-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16856005
|
Proteintech
28118-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MUC5B antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MUC5B. This antibody recognizes human antigen. The MUC5B antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| MUC5B | |
| Immunohistochemistry, ELISA | |
| Unconjugated | |
| Human | |
| IgG |
| 333 μg/mL | |
| Polyclonal | |
| Rabbit | |
| 727897 | |
| Primary |