missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC6 Antibody (MUC6/1553R), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08352F
This item is not returnable.
View return policy
Description
MUC6 Monoclonal antibody specifically detects MUC6 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MUC6 | |
| Monoclonal | |
| FITC | |
| gastric mucin, Gastric mucin-6, MUC-6, mucin 6, gastric, mucin 6, oligomeric mucus/gel-forming, mucin-6 | |
| Recombinant full-length human MUC6 protein (Uniprot: Q6W4X9) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| MUC6/1553R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 4588 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction