missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC6 Antibody (MUC6/916), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47799JF549
This item is not returnable.
View return policy
Description
MUC6 Monoclonal antibody specifically detects MUC6 in Human, Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MUC6 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| gastric mucin, Gastric mucin-6, MUC-6, mucin 6, gastric, mucin 6, oligomeric mucus/gel-forming, mucin-6 | |
| Recombinant fragment covering tandem repeats of human MUC6 protein (exact sequence is proprietary) (Uniprot: Q6W4X9) | |
| 0.1 mL | |
| Primary | |
| Human, Rat (Negative) | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MUC6/916 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 4588 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction