missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mucobromic Acid, TRC
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Specifications
Specifications
| Chemical Name or Material | Mucobromic Acid |
| CAS | 488-11-9 |
| Molecular Formula | C4H2Br2O3 |
| Quantity | 100 g |
| Synonym | (Z)-2,3-Dibromo-4-oxo-2-butenoic Acid, Dibromomalealdehydic Acid, Bromomucic Acid |
| InChI Formula | InChI=1S/C4H2Br2O3/c5-2(1-7)3(6)4(8)9/h1H,(H,8,9)/b3-2- |
| SMILES | Br/C(C=O)=C(Br)/C(O)=O-O=C1C(Br)=C(Br)C(O)O1 |
| IUPAC Name | (Z)-2,3-dibromo-4-oxobut-2-enoic acid |
| Molecular Weight (g/mol) | 257.86 |
| Formula Weight | 255.87 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction