Learn More
Hartmann Paul™ Mullro
Specifications
Specifications
| Product Type | Gauze Bandage Roll |
| Length (Metric) | 5000 mm |
| Width (Metric) | 800 mm |
| Quantity | 1 Ea. |
| Dimensions (L x W) | 5000 x 800 mm |
| For Use With (Application) | As a universal dressing , in First aid |
| Absorbancy | Yes |
| Certifications/Compliance | EN 14079-VM 20 |