Learn More
Multi-element Ionic Chromatography Cation Standard solution, H2O matrix, Fisher chemical™
Product Code. p-7083727 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
Product Code. 12914375
Brand: Fisher Chemical CM 1772
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Multi-element Ionic Chromatography Cation Standard solution containing 4 components; 10mg/kg each of Lithium (Li+), Sodium (Na+), Potassium (K+), Calcium (Ca2+) for Ion Chromatography H2O matrix
Specifications
Chemical Name or Material Four Components Solution
Packaging Bottle
Analyte or Component Names Lithium, Sodium, Potassium, Calcium
Physical Form Liquid
Concentration 10mg/Kg of each component
Type Multi-component Cationic Standard Solution
Element Li, Na, K, Ca
Solvent or Matrix Water
Quantity 100 mL