Learn More
Multi-Element Organometallic Oil Standards 23 Element, 300μg/g; SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep CL-MEM-SET
Additional Details : Weight : 0.09000kg
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ag | 300μg/g |
Al | 300μg/g |
B | 300μg/g |
Ba | 300μg/g |
Ca | 300μg/g |
Cd | 300μg/g |
Cr | 300μg/g |
Cu | 300μg/g |
Fe | 300μg/g |
K | 300μg/g |
Mg | 300μg/g |
Mn | 300μg/g |
Mo | 300μg/g |
Na | 300μg/g |
Ni | 300μg/g |
P | 300μg/g |
Pb | 300μg/g |
Sb | 300μg/g |
Si | 300μg/g |
Sn | 300μg/g |
Ti | 300μg/g |
V | 300μg/g |
Zn | 300μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
23 Element Oil Standard | |
50 g | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
Base Oil 75 |
Triple-leached LDPE bottle | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Al,Ba,B |
Product intended for laboratory use only