Learn More
Shamrock™ Multi-Purpose Blank Tyvek Labels
Provides extra strength, adhesion and permanent marking for your laboratory needs. Shamrock™ Multi-Purpose Blank Tyvek Labels won't discolor, crack or tear, adhere to almost any surface and are available in a range of sizes.
€ 157.00 - € 270.00
Specifications
Adhesive Type | Permanent |
---|---|
Color | White |
Material | Tyvek |
Product Type | Multi-Purpose Blank Tyvek Label |
For Use With (Application) | For Unique and demanding applications |
Product Code | Brand | Size (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size (Metric) | Price | Quantity & Availability | |||||
11822651
|
Shamrock™
STV-4 |
38 x 76 mm |
€ 202.00
Pack of 1000 |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11832651
|
Shamrock™
STV-5 |
51 x 100 mm |
€ 270.00
Pack of 1000 |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11892641
|
Shamrock™
STV-1 |
20 x 22 mm |
€ 157.00
Pack of 1000 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11802651
|
Shamrock™
STV-2 |
20 x 38 mm |
€ 157.00
Pack of 1000 |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Tear-proof material, long-lasting adhesive
- Does not discolor over time
- Permanent marking with ballpoint pen, paper pencil and felt marker
- Labels pre-cut on roll
Specifications
Permanent | |
Tyvek | |
For Unique and demanding applications |
White | |
Multi-Purpose Blank Tyvek Label |