Learn More
ZVG Zellstoff Vertrieb™ Multiclean Plus Cloth Rolls
Product Code. 10639342
Click to view available options
Ply:
2-ply
3-ply
Width (Metric):
240 mm
380 mm
Unit Size:
1 roll
2 rolls
Product Code. 10639342
Brand: ZVG Zellstoff Vertrieb™ 1133801
Edge
Clean using ZVG Zellstoff Vertrieb™ Multiclean Plus Cloth Rolls for a high degree of efficiency and safety A new manufacturing process makes these cloths even stronger and more absorbent.
  • Blue
  • 2-ply and 3-ply
  • High volume
Specifications
Product Type Multiclean Plus Cloth Roll
Length (Metric) 380 mm
Width (Metric) 380 mm
Color Blue
Ply 3-ply