Learn More
Hanna Instruments™ Multiparameter Benchtop Photometer
Specifications
Specifications
| Product Type | Multiparameter Benchtop Photometer |
| Display Type | LCD |
| Connection Type | USB |
| Height (Metric) | 110 mm |
| Length (Metric) | 235 mm |
| Light Source | Up to 5 tungsten lamps with different narrow band interference filters |
| Weight (Metric) | 0.9 kg |
| Width (Metric) | 200 mm |