Learn More
ZVG Zellstoff Vertrieb™ ZetBox™ Laboratory and Cosmetic Towels
Product Code. 10341582
Click to view available options
Color:
White
Unit Size:
Pack of 30
Product Code. 10341582
Brand: ZVG Zellstoff Vertrieb™ 1651500_OLD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Remove each robust towel individually thanks to the rounded opening of the robust box.After opening the box of ZVG Zellstoff Vertrieb™ ZetBox™ Laboratory and Cosmetic Towels, two plastic lids ensure that after each removal of a single cloth, the remaining cloths are protected from external influences.
  • Double layered
  • ]Ultrasoft
  • Protected rounded box opening
  • Ultra-absorbent
Specifications
Product Type Multi-purpose Towels
Length (Metric) 21 cm
Width (Metric) 22 cm
Color White
Disposable Single-use
Dimensions (L x W) 21 x 22 cm
Packaging Quantity 30 Boxes
Ply 2-ply