missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Multiskan Sky Microplate Spectrophotometer

Multiskan Sky Microplate Spectrophotometer is an absorbance microplate reader designed for the ultimate user experience. By introducing an attractive, touchscreen based user interface combined with multiple connectivity options, we offer refined photometry and bring unmatched versatility to academic, biotech, and pharmaceutical laboratories.

Brand:  Thermo Scientific™ 51119600DP

Additional Details : Weight : 13.80000kg

 View more versions of this product

Product Code. 15738147

  • € 13930.00 / Each


Estimated Shipment
13-01-2021
Add to basket

Description

Description

  • Monochromator-based optical system for free selection of wavelengths from 200 nm to 1000 nm
  • Compatible with 96- and 384-well microplates with and without lids or cuvettes
  • Reads μDrop™ Plate for micro-volume DNA/RNA and protein analysis
  • Performs onboard shaking and incubation for temperature-critical or cell-based assays
  • Allows kinetic, spectral, and endpoint measurements for a variety of spectrophotometer applications
  • Fast operation: full spectrum of a well is done in less than 10 seconds and a full 96-well microplate is read in 6 seconds
  • Touchscreen models have an easy-to-use interface for stand-alone use and includes ready-made protocols for nucleic acid and protein quantification
  • Setting up of demanding assays is easy via the powerful and intuitive SkanIt™ Software
  • Thermo Fisher Connect cloud-based tools allow you to securely store, access, share, and manage your photometric data remotely (touchscreen models)
  • Multiple languages available on all models; choose between English, German, French, Spanish, Italian, Portuguese, Russian, Chinese, and Japanese
  • Automation compatibility for integration with SkanIt Software automation interface
Specifications

Specifications

Multiskan Sky with touchscreen and μDrop Plate
Xenon flash lamp
No
200-1000 nm
0–2.5 Abs, 2% @ 450 nm
1.0% + 0.003 Abs (0–2.0 Abs), 2.0% (2.0–2.5 Abs) @ 450 nm
Linear shaking
10 s from 200 to 1000 nm
English, German, French, Spanish, Italian, Portuguese, Russian, Chinese, Japanese
11.6 in.
23.8 lbs
Automation
1 year
96- and 384-well formats and μDrop Plate
Monochromator
< 2.5 nm
1 nm
Ambient + 2°C to 45°C
96-well microplate: 6 seconds (fast mode)384-well microplate: 10 seconds (fast mode)
Touchscreen display and SkanIt PC software
17.5 in.
10.4 in.
µDrop plate
Videos
SDS
Documents
Warranty