missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUM1 Monoclonal Antibody (OTI1B2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804374
This item is not returnable.
View return policy
Description
Mutated melanoma-associated antigen 1 (MUM1) is a 78 kDa protein encoded by the MUM1 gene. MUM1 is expressed in a subset of B cells in the light zone of the germinal center, plasma cells, activated T cells and a wide spectrum of related hematolymphoid neoplasms. Antibodies to MUM1 are useful in the sub classification of lymphoid maligcies.Specifications
| MUM1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q2TAK8 | |
| MUM1 | |
| Human recombit protein fragment corresponding to amino acids 1-270 of human MUM1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1B2 | |
| Unconjugated | |
| MUM1 | |
| EXPAND1, HSPC211, MUM-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 84939 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction