missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mumps Virus NP Monoclonal Antibody (6008)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA547563

Product Code. 17865127

  • € 491.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specificity: Specific for the nucleoprotein of Mumps virus (see below). Does not cross react with: Measles virus, Respiratory Syncytial virus, Parainfluenza 1, 2 or 3 viruses. Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.

Mumps rubulavirus is the causative agent of mumps. The signs of mumps include swelling of the parotid glands, salivary glands and other epithelial tissues. Symptoms of mumps are fatigue, body aches, headache, loss of appetite, low grade fever, swelling of the salivary glands. Mumps can also result in muscle pain, deafness, meningitis, pancreatitis, swelling of testicles or ovaries, and death. Most people who contract mumps show symptoms of the virus, however there are few who show no or very few symptoms. Natural infection is currently restricted to humans and the virus is transmitted by direct contact, droplet spread, or contaminated objects.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mumps Virus NP
Monoclonal
100 μg/mL
PBS with 0.1% sodium azide; pH 7.2
Mouse
Protein A
RUO
Virus
Antibody
IgG2b
ELISA, Immunocytochemistry
6008
Unconjugated
mumps virus nuclear
Proprietary sequence.
100 μg
Primary
4°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.