missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CaMKII Pan Recombinant Rabbit Monoclonal Antibody (23GB5920), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552951
This item is not returnable.
View return policy
Description
The Ca2+/calmodulin-dependent protein kinases (CaM kinases) comprise a structurally related subfamily of serine/threonine kinases which include CaMKI, CaMKII and CaMKIV. CaMKII is an ubiquitously expressed serine/threonine protein kinase that is activated by Ca2+/calmodulin (CaM) binding and autophosphorylation and has been implicated in regulation of the cell cycle and transcription. It is involved in dendritic spine and synapse formation, neuronal plasticity and regulation of sarcoplasmic reticulum Ca2+ transport in skeletal muscle. In neurons, plays an essential structural role in the reorganization of the actin cytoskeleton during plasticity by binding and bundling actin filaments in a kinase-independent manner.
Specifications
| CaMKII Pan | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Camk2b | |
| [d]-CaMKII; 2810011D23Rik; 8030469K03Rik; alpha CaM kinase II; alpha-CaMKII; Ca++/calmodulin-dependent protein kinase 2 delta subunit; Ca++/calmodulin-dependent protein kinase II beta subunit; Ca++/calmodulin-dependent protein kinase II delta subunit; Ca++/calmodulin-dependent protein kinase II, delta subunit; Ca2+/calmodulin-dependent protein kinase II; Ca2+/calmodulin-dependent protein kinase II alpha; calcium/calmodulin dependent protein kinase II alpha; calcium/calmodulin dependent protein kinase II beta; calcium/calmodulin dependent protein kinase II delta; calcium/calmodulin dependent protein kinase II gamma; calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II beta; calcium/calmodulin-dependent protein kinase (CaM kinase) II delta; calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma; calcium/calmodulin-dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase II alpha subunit; calcium/calmodulin-dependent protein kinase II alpha-B subunit; calcium/calmodulin-dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase II delta; calcium/calmodulin-dependent protein kinase II gamma; calcium/calmodulin-dependent protein kinase II, beta; calcium/calmodulin-dependent protein kinase II, delta; calcium/calmodulin-dependent protein kinase type II alpha chain; calcium/calmodulin-dependent protein kinase type II beta chain; calcium/calmodulin-dependent protein kinase type II delta chain; calcium/calmodulin-dependent protein kinase type II gamma chain; calcium/calmodulin-dependent protein kinase type II subunit alpha; Calcium/calmodulin-dependent protein kinase type II subunit beta; calcium/calmodulin-dependent protein kinase type II subunit delta; Calcium/calmodulin-dependent protein kinase type II subunit gamma; CaM kinase II alpha subunit; CaM kinase II beta subunit; CaM kinase II delta subunit; caM kinase II subunit alpha; CaM kinase II subunit beta; caM kinase II subunit delta; CaM kinase II subunit gamma; CAM2; CAMK; CaMK II; CAMK1; Camk2; Camk2a; Camk2b; Camk2d; Camk2g; CAMKA; CAMKB; CAMKD; CAMKG; Camki; CaMKII; CAMK-II; CaMK-II alpha subunit; CaMK-II delta subunit; caMK-II subunit alpha; caMK-II subunit beta; caMK-II subunit delta; caMK-II subunit gamma; CaMKIINalpha; CaM-kinase II alpha chain; CaM-kinase II beta chain; CaM-kinase II delta chain; caM-kinase II gamma chain; Ck2b; KIAA0968; Kiaa4163; mKIAA0968; PK2CDD; PKCCD; proline rich calmodulin-dependent protein kinase; R74975; RATCAMKI | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 108058, 12322, 12323, 12325, 815, 816, 817, 818 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5920 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P11798, P28652, Q13554, Q13555, Q13557, Q6PHZ2, Q923T9, Q9UQM7 | |
| Camk2a, Camk2b, CAMK2D, Camk2g | |
| A synthesized peptide derived from human CaMKII (1-88AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction