missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUPP1 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-49975H
This item is not returnable.
View return policy
Description
MUPP1 Polyclonal antibody specifically detects MUPP1 in Human samples. It is validated for Western Blot
Specifications
| MUPP1 | |
| Polyclonal | |
| Western Blot | |
| DKFZp781P216, FLJ25909, FLJ90240, Multi-PDZ domain protein 1, multiple PDZ domain protein, MUPP1FLJ34626 | |
| The immunogen for this product maps to a region between residue 1125 and 1150 of human Multiple PDZ Domain Protein using the numbering given in entry NP_003820.2 (GeneID 8777). | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 8777 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction