missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MURF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554058
This item is not returnable.
View return policy
Description
Immunogen sequence: MEEPQMALYL QQAKELINKV GAMSKVELAG RPEPGYESME QFTVRVEHVA EMLRTIDFQP GASGEEEEVA PDGEEGSAGP EEERPDG Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 88%.
The protein encoded by this gene contains a RING finger motif and is highly similar to the ring finger proteins RNF28/MURF1 and RNF29/MURF2. In vitro studies demonstrated that this protein, RNF28, and RNF29 form heterodimers, which may be important for the regulation of titin kinase and microtubule-dependent signal pathways in striated muscles. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
| MURF3 | |
| Polyclonal | |
| Unconjugated | |
| TRIM54 | |
| 4930486E09Rik; 4930566I02Rik; MURF; MURF3; MuRF-3; Muscle-specific RING finger protein; Muscle-specific RING finger protein 3; muscle-specific RING-finger protein; muscle-specific RING-finger protein 3; OTTHUMP00000122598; ring finger protein 30; Rnf30; TRIM54; tripartite motif containing 54; tripartite motif-containing 54; tripartite motif-containing protein 54 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57159 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BYV2 | |
| TRIM54 | |
| Recombinant protein corresponding to Human MURF3. Recombinant protein control fragment (Product #RP-92301). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction