Learn More
Murine Hepatitis Virus A59 nsp9 Antibody (2C6.H1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-21671CL1
This item is not returnable.
View return policy
Description
Murine Hepatitis Virus A59 nsp9 Monoclonal antibody specifically detects Murine Hepatitis Virus A59 nsp9 in Virus samples. It is validated for Western Blot,Dot Blot,Immunocytochemistry/Immunofluorescence
Specifications
| Murine Hepatitis Virus A59 nsp9 | |
| Monoclonal | |
| CoraFluor 1 | |
| 2'-O-MT, 3' to 5' exonuclease, 3C-like proteinase, HEL1, leader protein, NendoU, nsp10, nsp11, NSP2, nsp4, nsp6, nsp7, nsp8, nsp9, ORF1a polyprotein, ORF1ab polyprotein, PLpro, polyprotein pp1a, polyprotein pp1ab, RNA-dependent RNA polymerase | |
| This Murine Hepatitis Virus A59 nsp9 Antibody (2C6.H1) was produced in mice by repeated immunizations with E.coli derived full-length Murine Hepatitis Virus A59 nsp9 protein. This protein is part of the viral replicase polyprotein. (Uniprot: P0C6V0) | |
| 0.1 mL | |
| Primary | |
| Virus | |
| Purified |
| Western Blot, Dot Blot, Immunocytochemistry/Immunofluorescence | |
| 2C6.H1 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 1489749 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b κ |