missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mus81 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97261IR
This item is not returnable.
View return policy
Description
Mus81 Polyclonal antibody specifically detects Mus81 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Mus81 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| crossover junction endonuclease MUS81, EC 3.1.22, EC 3.1.22.-, FLJ21012, FLJ44872, MUS81 endonuclease homolog (S. cerevisiae), MUS81 endonuclease homolog (yeast), SLX3, SLX3 structure-specific endonuclease subunit homolog | |
| Produced in rabbits immunized with E. coli-derived Human Mus81 fragment. | |
| 0.1 mL | |
| DNA Repair, Homologous Recombination | |
| 80198 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction