Learn More
Mus81 Antibody (mta30 2g10/3), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-2064F
Product Code. 18025176
  • ONLINE EXCLUSIVE PRICE
    € 537.00
    € 507.15 / 0.10mL
    Save € 29.85
    5% Off
Description
Mus81 Monoclonal specifically detects Mus81 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mus81
Monoclonal
FITC
PBS with 0.05% Sodium Azide
MUS81
Human MUS81
0.1 mL
DNA Repair, Homologous Recombination
80198
Store at 4C in the dark.
IgG2a
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
mta30 2g10/3
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
crossover junction endonuclease MUS81, EC 3.1.22, EC 3.1.22.-, FLJ21012, FLJ44872, MUS81 endonuclease homolog (S. cerevisiae), MUS81 endonuclease homolog (yeast), SLX3, SLX3 structure-specific endonuclease subunit homolog
Mouse
Protein A purified