Learn More
Mus81 Antibody (MTA30 2G10/3), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-2064MFV450
Product Code. 18666749
  • € 619.00 / 0.10mL
Description
Mus81 Monoclonal antibody specifically detects Mus81 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mus81
Monoclonal
mFluor Violet 450 SE
Store at 4°C in the dark.
Flow Cytometry
MTA30 2G10/3