Learn More
Mus81 Antibody (MTA30 2G10/3), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2064MFV450
This item is not returnable.
View return policy
Description
Mus81 Monoclonal antibody specifically detects Mus81 in Human samples. It is validated for Flow Cytometry
Specifications
| Mus81 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| MTA30 2G10/3 | |
| Primary | |
| IgG2a |