Learn More
Musashi-2 Antibody (960121), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB3255APCCY7
Product Code. 30032648
  • € 650.00 / 0.10mL
Description
Musashi-2 Monoclonal antibody specifically detects Musashi-2 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Musashi-2
Monoclonal
APC-Cyanine7
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry
960121
Flow Cytometry
FLJ36569, MGC3245, MSI2H, musashi homolog 2 (Drosophila), musashi-2, RNA-binding protein Musashi homolog 2
E. coli-derived recombinant human Musashi-2, Met1-His328, Accession # Q96DH6
0.1 mL
Neuronal Cell Markers
124540
Store at 4C in the dark. Do not freeze.
IgG2b