missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Musashi-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30398
This item is not returnable.
View return policy
Description
Musashi-2 Polyclonal antibody specifically detects Musashi-2 in Human samples. It is validated for Immunoprecipitation
Specifications
| Musashi-2 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| FLJ36569, MGC3245, MSI2H, musashi homolog 2 (Drosophila), musashi-2, RNA-binding protein Musashi homolog 2 | |
| between 250 and 300 | |
| 100 μg | |
| Neuronal Cell Markers | |
| 124540 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction