missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Musashi-2 Antibody (OTI2F10), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72836APC
This item is not returnable.
View return policy
Description
Musashi-2 Monoclonal antibody specifically detects Musashi-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Musashi-2 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| OTI2F10 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| FLJ36569, MGC3245, MSI2H, musashi homolog 2 (Drosophila), musashi-2, RNA-binding protein Musashi homolog 2 | |
| Full length human recombinant protein of human MSI2(NP_620412) produced in HEK293T cell. | |
| 0.1 mL | |
| Neuronal Cell Markers | |
| 124540 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction