missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Muscarinic Acetylcholine Receptor M2/CHRM2 Antibody (585713), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne FAB6377M
This item is not returnable.
View return policy
Description
Muscarinic Acetylcholine Receptor M2/CHRM2 Monoclonal antibody specifically detects Muscarinic Acetylcholine Receptor M2/CHRM2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry
Specifications
| Muscarinic Acetylcholine Receptor M2/CHRM2 | |
| Monoclonal | |
| DyLight 594 | |
| 7TM receptor, cholinergic receptor, muscarinic 2, FLJ43243, HM2, MGC120006, MGC120007, muscarinic acetylcholine receptor M2, muscarinic M2 receptor | |
| NS0 mouse myeloma cell line transfected with human CHRM2, Accession # P08172 | |
| 0.1 mL | |
| GPCR, Neuroscience | |
| 1129 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry | |
| 585713 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction