missing translation for 'onlineSavingsMsg'
Learn More

Muscarinic Acetylcholine Receptor M3/CHRM3 Antibody (SR2111), Novus Biologicals™

Rabbit Monoclonal Antibody

€ 373.00 - € 483.00

Specifications

Antigen Muscarinic Acetylcholine Receptor M3/CHRM3
Clone SR2111
Dilution Western Blot 1:500-1:1000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18655964
View Documents
Novus Biologicals
NBP3-21828-100ul
100 μg
€ 483.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18680815
View Documents
Novus Biologicals
NBP3-21828-50ul
50 μg
€ 373.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Muscarinic Acetylcholine Receptor M3/CHRM3 Monoclonal antibody specifically detects Muscarinic Acetylcholine Receptor M3/CHRM3 in Human samples. It is validated for Western Blot
Specifications

Specifications

Muscarinic Acetylcholine Receptor M3/CHRM3
Western Blot 1:500-1:1000
Monoclonal
Purified
RUO
Human
cholinergic receptor, muscarinic 3, HM3, m3 muscarinic receptor, muscarinic acetylcholine receptor M3
A synthesized peptide derived from human Muscarinic Acetylcholine Receptor M3/CHRM3 (Uniprot #: P20309)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR2111
Western Blot
Unconjugated
Rabbit
GPCR
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
1131
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.