missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Muscle Phosphofructokinase/PFKM/PFK-1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32169AF647
This item is not returnable.
View return policy
Description
Muscle Phosphofructokinase/PFKM/PFK-1 Polyclonal specifically detects Muscle Phosphofructokinase/PFKM/PFK-1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Muscle Phosphofructokinase/PFKM/PFK-1 | |
| Polyclonal | |
| Western Blot | |
| 6-phosphofructokinase, muscle type, EC 2.7.1,6-phosphofructo-1-kinase, EC 2.7.1.11, GSD7, MGC8699, PFK1, PFK-1, PFKA, PFK-A, PFKX, Phosphofructo-1-kinase isozyme A, Phosphofructokinase 1, phosphofructokinase, muscle, phosphofructokinase, polypeptide X, phosphofructokinase-M, phosphohexokinase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PFKM | |
| The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Phosphofructokinase, Muscle using the numbering given in entry NP_000280.1 (GeneID 5213). | |
| 100 μL | |
| Lipid and Metabolism | |
| 5213 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction