missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Muscle Phosphofructokinase/PFKM/PFK-1 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32170DL594
This item is not returnable.
View return policy
Description
Muscle Phosphofructokinase/PFKM/PFK-1 Polyclonal antibody specifically detects Muscle Phosphofructokinase/PFKM/PFK-1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Muscle Phosphofructokinase/PFKM/PFK-1 | |
| Polyclonal | |
| Western Blot | |
| 6-phosphofructokinase, muscle type, EC 2.7.1,6-phosphofructo-1-kinase, EC 2.7.1.11, GSD7, MGC8699, PFK1, PFK-1, PFKA, PFK-A, PFKX, Phosphofructo-1-kinase isozyme A, Phosphofructokinase 1, phosphofructokinase, muscle, phosphofructokinase, polypeptide X, phosphofructokinase-M, phosphohexokinase | |
| The immunogen this antibody was made to, maps to a region between residue 730 to 780 of human Phosphofructokinase, Muscle using the numbering given in entry NP_000280.1 (GeneID 5213). | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 5213 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction