Learn More
Muscone, ≥95%, MP Biomedicals™
Product Code. 11461391 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 11461391
Brand: MP Biomedicals 0215918680
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Muscone, originally collected from male musk deer ventral glands, is a potent antiinflammatory agent that has been reported to reduce proinflammatory cytokines in vitro
TRUSTED_SUSTAINABILITY
Specifications
CAS 541-91-3
Color Colorless
Density 0.84g/mL
Assay Percent Range ≥95%
Molecular Formula C16H30O
MDL Number MFCD00211114
Quantity 100 mg
Synonym muscone, 3-methylcyclopentadecanone, muskone, methylexaltone, moschus ketone, cyclopentadecanone, 3-methyl, 3-methyl-1-cyclopentadecanone, 5-methyl-1-cyclopentadecanone, fema no. 3434, 3-methyl-cyclopentadecanone
InChI Key ALHUZKCOMYUFRB-UHFFFAOYNA-N
SMILES CC1CCCCCCCCCCCCC(=O)C1
IUPAC Name 3-methylcyclopentadecan-1-one
Molecular Weight (g/mol) 238.42
PubChem CID 10947
Formula Weight 238.43
Physical Form Liquid
Chemical Name or Material Muscone