Learn More
Muskelin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565312
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: ETVYLFGGWDG TQDLADFWAY SVKENQWTCI SRDTEKENGP SARSCHKMCI DIQRRQIYTL GRYLDSSVRN SKSLKSDFYR YDIDTNTWML LSEDTAADGG PKLVFDHQMC MDSE Highest antigen sequence identity to the following orthologs - mouse 100%, rat 98%.
Muskelin is an intracellular protein that acts as a mediator of cell spreading and cytoskeletal responses to the extracellular matrix component thrombospondin I.Specifications
Muskelin | |
Polyclonal | |
Unconjugated | |
MKLN1 | |
A130067F06Rik; AU015903; BB154892; cytoplasmic mediator; fc60a07; LOW QUALITY PROTEIN: muskelin; Mkln1; mkln1 protein; Msk; Muskelin; muskelin 1; muskelin 1, intracellular mediator containing kelch motifs; TWA2; wu:fc60a07 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4289 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UL63 | |
MKLN1 | |
Recombinant Human MNK1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only