Learn More
MUT Mouse anti-Human, Clone: OTI2A8, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA506883S
Encompass_Preferred
Product Code. 17506512
  • € 242.00 / 30µL
Description
MUT Monoclonal antibody specifically detects MUT in Human samples. It is validated for Immunocytochemistry, Western Blot
Specifications
Specifications
MUT
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P22033
Mouse
Affinity chromatography
RUO
4594
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunocytochemistry, Western Blot
OTI2A8
Unconjugated
MMUT
D230010K02Rik; Mcm; methylmalonyl CoA mutase; methylmalonyl Coenzyme A mutase; methylmalonyl-CoA isomerase; methylmalonyl-CoA mutase; methylmalonyl-CoA mutase, mitochondrial; methylmalonyl-Coenzyme A mutase; MMUT; MUT; mutant methylmalonyl CoA mutase; truncated methylmalonyl CoA mutase
Full length human recombinant protein of human MUT produced in HEK293T cell.