missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ MUT Recombinant Rabbit Monoclonal Antibody (23GB4550), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553764
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
The product of this gene is a Golgi enzyme catalyzing an essential step in the conversion of oligomannose to complex N-glycans. The enzyme has the typical glycosyltransferase domains: a short N-terminal cytoplasmic domain, a hydrophobic non-cleavable signal-anchor domain, and a C-terminal catalytic domain. Mutations in this gene may lead to carbohydrate-deficient glycoprotein syndrome, type II. The coding region of this gene is intronless. Transcript variants with a spliced 5' UTR may exist, but their biological validity has not been determined.
Tekniske data
| MUT | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MMUT | |
| D230010K02Rik; Mcm; methylmalonyl CoA mutase; methylmalonyl Coenzyme A mutase; methylmalonyl-CoA isomerase; methylmalonyl-CoA mutase; methylmalonyl-CoA mutase, mitochondrial; methylmalonyl-Coenzyme A mutase; MMUT; MUT; mutant methylmalonyl CoA mutase; truncated methylmalonyl CoA mutase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17850, 4594, 688517 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4550 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P16332, P22033 | |
| MMUT | |
| A synthesized peptide derived from human MUTA (550-580AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion