missing translation for 'onlineSavingsMsg'
Learn More

Mutant IDH1-IN-2, MedChemExpress

Product Code. 16440078
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16440078 5 mg 5mg
16407608 10 mg 10mg
16403627 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16440078 Supplier Medchem Express Supplier No. HY18717

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mutant IDH1-IN-2 is a inhibitor of mutant Isocitrate dehydrogenase (IDH) proteins, with IC50 of in LS-MS biochemical assay, IC50 of 16.6 nM in Fluorescence biochemical assay.

Mutant IDH1-IN-2 is a inhibitor of mutant Isocitrate dehydrogenase (IDH) proteins, with IC50 of in LS-MS biochemical assay, IC50 of 16.6 nM in Fluorescence biochemical assay.

Specifications

Chemical Name or Material Mutant IDH1-IN-2
CAS 1429176-69-1
Color White
Purity Grade Notes Research
Molecular Formula C24H31F2N5O2
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (108.81 mM; Need ultrasonic)
SMILES O=C1OC[C@H](C(C)C)N1C2=NC(N[C@H](C3=CC=C(CN4CCC(F)(F)CC4)C=C3)C)=NC=C2
Molecular Weight (g/mol) 459.53
Formula Weight 459.53
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.