missing translation for 'onlineSavingsMsg'
Learn More

Mutated EGFR-IN-1, MedChemExpress

Product Code. 16444127
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16444127

Brand: Medchem Express HY78869

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mutated EGFR-IN-1 (Osimertinib analog) is a useful intermediate for the inhibitors design for mutated EGFR, such as L858R EGFR, Exonl9 deletion activating mutant and T790M resistance mutant.

Mutated EGFR-IN-1 (Osimertinib analog) is a useful intermediate for the inhibitors design for mutated EGFR, such as L858R EGFR, Exonl9 deletion activating mutant and T790M resistance mutant.

Specifications

Chemical Name or Material Mutated EGFR-IN-1
CAS 1421372-66-8
Color Pale Purple
Purity Grade Notes Research
Molecular Formula C25H31N7O
Quantity 50 mg
Synonym Osimertinib analog
Solubility Information DMSO : ≥ 75 mg/mL (168.33 mM)
SMILES NC1=CC(NC2=NC=CC(C3=CN(C)C4=C3C=CC=C4)=N2)=C(OC)C=C1N(CCN(C)C)C
Molecular Weight (g/mol) 445.56
Formula Weight 445.56
Percent Purity 98.59%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.