missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MVD Antibody (SR1738), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22438-50ul
This item is not returnable.
View return policy
Description
MVD Monoclonal antibody specifically detects MVD in Human samples. It is validated for Western Blot
Specifications
| MVD | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| SR1738 | |
| Western Blot 1:500-1:2000 | |
| EC 4.1.1.33, FP17780, MDDase, mevalonate (diphospho) decarboxylase, Mevalonate (diphospho)decarboxylase, Mevalonate pyrophosphate decarboxylase, MPDdiphosphomevalonate decarboxylase | |
| A synthesized peptide derived from human MVD (Uniprot #: P53602) | |
| 50 μg | |
| Lipid and Metabolism | |
| 4597 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction