Learn More
MVK Mouse anti-Human, Unconjugated, Clone: 2F1D3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67820-1-IG-150UL
 View more versions of this product
Product Code. 17531426
  • € 410.00 / 150µL
Description
The MVK antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human MVK. This antibody recognizes human antigen. The MVK antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
MVK
Monoclonal
Unconjugated
150 μL
4598
IgG2b
Western Blot, ELISA
2F1D3