missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MVP Antibody (1032), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47807PE
This item is not returnable.
View return policy
Description
MVP Monoclonal antibody specifically detects MVP in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Specifications
| MVP | |
| Monoclonal | |
| PE | |
| LRPVAULT1, Lung resistance-related protein, major vault protein | |
| proteins precipitated from human breast cancer MCF-7 cells | |
| 0.1 mL | |
| Cancer | |
| 9961 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 1032 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction