missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MVP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30548
This item is not returnable.
View return policy
Description
MVP Polyclonal antibody specifically detects MVP in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| MVP | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| LRPVAULT1, Lung resistance-related protein, major vault protein | |
| Between 843 and 893 | |
| 100 μg | |
| Cancer, Signal Transduction | |
| 9961 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction