Learn More
MX1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556590
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: IEDIRAEQER EGEKLIRLHF QMEQIVYCQD QVYRGALQKV REKELEEEKK KKSWDFGAFQ SSSATDSS Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 60%.
This gene encodes a guanosine triphosphate (GTP)-metabolizing protein that participates in the cellular antiviral response. The encoded protein is induced by type I and type II interferons and antagonizes the replication process of several different RNA and DNA viruses. There is a related gene located adjacent to this gene on chromosome 21, and there are multiple pseudogenes located in a cluster on chromosome 4. Alternative splicing results in multiple transcript variants.Specifications
MX1 | |
Polyclonal | |
Unconjugated | |
Mx1 | |
AI893580; GTP-binding protein; IFI78; IFI-78K; Influenza resistance protein; Interferon-induced GTP-binding protein Mx1; Interferon-induced GTP-binding protein Mx1, N-terminally processed; Interferon-induced protein p78; interferon-inducible myxovirus resistance-1 protein; interferon-inducible protein p78; interferon-regulated resistance GTP-binding protein MxA; Mx; MX dynamin like GTPase 1; MX dynamin-like GTPase 1; Mx1; Mx-1; Mx1B; Mx1-b; MxA; Myxoma resistance protein 1; myxovirus (influenza virus) resistance 1; myxovirus (influenza virus) resistance 1, interferon-inducible protein p78; myxovirus (influenza) resistance 1 polypeptide; myxovirus resistance protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4599 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P20591 | |
Mx1 | |
Recombinant protein corresponding to Human MX1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only