missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MxA/Mx1 Antibody (MX1/7529), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26822-20ug
This item is not returnable.
View return policy
Description
MxA/Mx1 Monoclonal antibody specifically detects MxA/Mx1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MxA/Mx1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| human interferon-regulated resistance GTP-binding protein MXA10IFI-78Khomolog of murine (interferon-inducibleprotein p78), myxovirus (influenza virus) resistance 1, interferon-inducible protein p78(mouse) | |
| Recombinant fragment (around aa400-592) of human MxA/Mx1 protein (exact sequence is proprietary) | |
| 20 μg | |
| Apoptosis | |
| 4599 | |
| Store at 4°C. | |
| IgG2 κ |
| Immunohistochemistry (Paraffin) | |
| MX1/7529 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction