Learn More
MxA/Mx1 Antibody (OTI2G12), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-72838AF488
Product Code. 18164686
  • ONLINE EXCLUSIVE PRICE
    € 807.00
    € 763.35 / 0.10mL
    Save € 43.65
    5% Off
Description
MxA/Mx1 Monoclonal specifically detects MxA/Mx1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MxA/Mx1
Monoclonal
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
MX1
Full length recombinant protein of human MxA/Mx1 (NP_002453) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse
IgG1
Western Blot, Immunocytochemistry, Immunofluorescence
OTI2G12
Western Blot, Immunocytochemistry/Immunofluorescence
human interferon-regulated resistance GTP-binding protein MXA10IFI-78Khomolog of murine (interferon-inducibleprotein p78), myxovirus (influenza virus) resistance 1, interferon-inducible protein p78(mouse)
Mouse
Affinity Purified
Apoptosis
4599.0
Store at 4°C in the dark.