missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MXD4 Monoclonal Antibody (OTI5E6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808855S
This item is not returnable.
View return policy
Description
MXD4 Monoclonal antibody specifically detects MXD4 in Human samples. It is validated for Western BlotSpecifications
| MXD4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NZI7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7342 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI5E6 | |
| Unconjugated | |
| UBP1 | |
| 2810410A03Rik; bHLHc12; Class C basic helix-loop-helix protein 12; LOW QUALITY PROTEIN: max dimerization protein 4; MAD4; Mad4 homolog; max dimerization protein 4; Max dimerizer 4; max-associated protein 4; Max-interacting transcriptional repressor MAD4; MST149; MSTP149; MXD4 | |
| Human recombinant protein fragment corresponding to amino acids 47-269 of human UBP1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction