missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mxi1 Antibody (PCRP-MXI1-1A3), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08353AF532
This item is not returnable.
View return policy
Description
Mxi1 Monoclonal antibody specifically detects Mxi1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array
Specifications
| Mxi1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| BHLHC11, Class C basic helix-loop-helix protein 11, MAX dimerization protein 2, MAX interacting protein 1, MAX interactor 1bHLHc11MAD2, max-interacting protein 1, Max-related transcription factor, MGC43220, MXD2, MXI | |
| Recombinant full-length human Mxi1 protein (Uniprot: P50539) | |
| 100 μg | |
| Cell Cycle and Replication | |
| 4601 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunofluorescence, Protein Array | |
| PCRP-MXI1-1A3 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction