missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MXRA5 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-46122
This item is not returnable.
View return policy
Description
MXRA5 Polyclonal specifically detects MXRA5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| MXRA5 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| Adhesion protein with leucine-rich repeats and immunoglobulin domains relatedto perlecan, Adlican, DKFZp564I1922, matrix-remodeling-associated protein 5, matrix-remodelling associated 5 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 25878 | |
| Human | |
| IgG |
| ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 ug/ml, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_056234.2 | |
| MXRA5 | |
| Peptide with sequence C-QAETTTLDQDHLET corresponding to internal region according to NP_056234.2. | |
| 0.1 mg | |
| Primary | |
| This antibody recognizes and epitope within the internal region. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction