missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYB, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00004602-D01.100uL
This item is not returnable.
View return policy
Description
Sequence: MARRPRHSIYSSDEDDEDFEMCDHDYDGLLPKSGKRHLGKTRWTREEDEKLKKLVEQNGTDDWKVIANYLPNRTDVQCQHRWQKVLNPELIKGPWTKEEDQRVIELVQKYGPKRWSVIAKHLKGRIGKQCRERWHNHLNPEVKKTSWTEEEDRIIYQAHKRLGNRWAEIAKLLPGRTDNAIKNHWNSTMRRKVEQEGYLQESSKASQPAVATSFQKNSHLMGFAQAPPTAQLPATGQPTVNNDYSYYHISEAQNVSSHVPYPVALHVNIVNVPQPAAAAIQRHYNDEDPEKEKRIKELELLLMSTENELKGQQVLPTQNHTCSYPGWHSTTIADHTRPHGDSAPVSCLGEHHSTPSLPADPGSLPEESASPARCMIVHQGTILDNVKNLLEFAETLQFIDSFLNTSSNHENSDLEMPSLTSTPLIGHKLTVTTPFHRDQTVKTQKENTVFRTPAIKRSILESSPRTPTPFKHALAAQEIKYGPLKMLPQTPSHLVEDLQDVIKQESDESGIVAEFQENGPPLLKKIKQEVESPTDKSGNFFCSHHWEGDSLNTQLFTQTSPVADAPNILTSSVLMAPASEDEDNVLKAFTVPKNRSLASPLQPCSSTWEPASCGKMEEQMTSSSQARKYVNAFSARTLVMSpecifications
| MYB | |
| Polyclonal | |
| No additive | |
| NM_005375.2 | |
| MYB | |
| MYB (NP_005366.2, 1 a.a. ∼ 640 a.a) full-length human protein. | |
| RUO | |
| 4602 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunoprecipitation | |
| Unconjugated | |
| v-myb myeloblastosis viral oncogene homolog (avian) | |
| Cmyb/c-myb/c-myb_CDS/efg | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction