missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYBBP1A Monoclonal Antibody (1702CT711.87.46)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537716
This item is not returnable.
View return policy
Description
MYBBP1A Monoclonal Antibody for Western Blot
The Myb family of transcription factors, which includes the structurally related A-, B-, and c-Myb genes, regulate differentiation, cellular growth and transactivating gene expression. c-Myb plays an essential role in controlling the proliferation and differentiation of hematopoietic cells, cellular levels of c-Myb decreasing as cells reach terminal differentiation. Myb-binding protein 1A (MYBBP1A) is a novel nuclear protein localized predominantly, but not exclusively, in nucleoli. Although initially isolated as a c-Myb-interacting protein, MYBBP1A is expressed ubiquitously, and associates with a number of different transcription factors. MYBBP1A associates with the aromatic hydrocarbon receptor (AhR), enhancing transactivation and increasing AhR-dependent gene expression. MYBBP1A is a powerful negative regulator of PPAR gamma coactivator 1 alpha (PGC-1 alpha) by p38 MAPK and also acts as a nucleocytoplasmic shuttling protein that utilizes CRM1-dependent and independent nuclear export pathways.
Specifications
| MYBBP1A | |
| Monoclonal | |
| 0.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9BQG0 | |
| MYBBP1A | |
| recombinant protein between 1104-1328 amino acids from human MYBBP1A. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| 1702CT711.87.46 | |
| Unconjugated | |
| MYBBP1A | |
| AL024407; AU019902; DBP; MYB binding protein 1a; MYB binding protein (P160) 1a; MYB binding protein 1a; myb-binding protein 1A; Myb-binding protein of 160 kDa; MYBBP1A; nuclear protein P160; P160; p160MBP; p53-activated protein-2; p67MBP; PAP2; PAR interacting protein; PAR-interacting protein; PIP | |
| Mouse | |
| Protein G | |
| RUO | |
| 10514 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction