missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYBPC1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-41157UV
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MYBPC1 Polyclonal antibody specifically detects MYBPC1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Especificaciones
| MYBPC1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 350 | |
| C-protein, skeletal muscle slow isoform, MYBPCC, MYBPCS, myosin binding protein C, slow type, myosin-binding protein C, slow-type, skeletal muscle C-protein, Slow MyBP-C, slow-type | |
| Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human MYBPC1. The immunogen is located within amino acids 160 - 210 of MYBPC1. Amino Acid Squence: DIRSAFKRSGEGQEDAGE | |
| 0.1 mL | |
| Cellular Signaling | |
| 4604 | |
| Store at 4°C in the dark. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido