missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYBPC1 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41119G
This item is not returnable.
View return policy
Description
MYBPC1 Polyclonal antibody specifically detects MYBPC1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MYBPC1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| C-protein, skeletal muscle slow isoform, MYBPCC, MYBPCS, myosin binding protein C, slow type, myosin-binding protein C, slow-type, skeletal muscle C-protein, Slow MyBP-C, slow-type | |
| Antibody was raised against a 12 amino acid synthetic peptide near the carboxy terminus of human MYBPC1. The immunogen is located within the last 50 amino acids of MYBPC1. Amino Acid Squence: LEGQQQSLHNKD | |
| 0.1 mL | |
| Cellular Signaling | |
| 4604 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction